Hexyl isobutyrate 98%

Supplier: Ambeed
Warning

A962724-100G A962724-25G A962724-500G
76777-054EA 10 USD
76777-054 76777-056 76777-058
Hexyl isobutyrate 98%
Hexyl isobutyrate

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
MDL Number: MFCD00027297
CAS Number: 2349-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR