(1-Methyl-1H-pyrazol-5-yl)methylamine 96%

Supplier: Ambeed
Warning

A961475-100MG A961475-1G A961475-250MG A961475-5G
76777-046EA 17 USD
76777-046 76777-048 76777-050 76777-052
(1-Methyl-1H-pyrazol-5-yl)methylamine 96%
(1-Methyl-1H-pyrazol-5-yl)methylamine

Precaución: for Research Use Only

Formula: C₅H₉N₃
MDL Number: MFCD03419808
CAS Number: 863548-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR