5-(2-Furyl)-1H-pyrazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A958136-100MG A958136-1G A958136-250MG
76777-036EA 29 USD
76777-036 77595-546 77878-757
5-(2-Furyl)-1H-pyrazole-3-carboxylic acid 97%
5-(2-Furyl)-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
MDL Number: MFCD05170057
CAS Number: 116153-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR