4-Phenylthiophene-2-carboxylic acid 99%

Supplier: Ambeed
Warning

A954076-100MG A954076-1G A954076-250MG
76777-030EA 13.8 USD
76777-030 76777-032 76777-034
4-Phenylthiophene-2-carboxylic acid 99%
4-Phenylthiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₈O₂S
MW: 204.25 g/mol
MDL Number: MFCD03408007
CAS Number: 21676-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR