5-Fluoro-1H-benzo[d]imidazol-2-amine 95%

Supplier: Ambeed
Warning

A952538-1G A952538-5G A952538-250MG
76777-026EA 147.47 USD
76777-026 76777-028 77627-776
5-Fluoro-1H-benzo[d]imidazol-2-amine 95%
5-Fluoro-1H-benzo[d]imidazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₆FN₃
MW: 151.14 g/mol
MDL Number: MFCD06659790
CAS Number: 30486-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR