DL-Methionine methylsulfonium chloride 98%

Supplier: Ambeed
Warning

Synonyms: Vitamin U, (3-Amino-3-carboxypropyl)dimethylsulphonium chloride

A948177-100G A948177-1G A948177-25G A948177-25MG A948177-500G A948177-1000G
76777-016EA 30 USD
76777-016 76777-018 76777-020 76777-022 76777-024 77599-290 77627-772
DL-Methionine methylsulfonium chloride 98%
DL-Methionine methylsulfonium chloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂S
MDL Number: MFCD00031700
CAS Number: 3493-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR