2-(Methylamino)-1H-purin-6(7H)-one 98%

Supplier: Ambeed
Warning

A935724-100MG A935724-250MG
76777-010EA 40.32 USD
76777-010 76777-012
2-(Methylamino)-1H-purin-6(7H)-one 98%
2-(Methylamino)-1H-purin-6(7H)-one

Precaución: for Research Use Only

Formula: C₆H₇N₅O
MW: 165.15 g/mol
MDL Number: MFCD00127896
CAS Number: 10030-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR