6-Chloro-2-methoxypyridin-3-amine 98%

Supplier: Ambeed
Warning

A933569-250MG A933569-5G
76777-006EA 19.1 USD
76777-006 76777-008
6-Chloro-2-methoxypyridin-3-amine 98%
6-Chloro-2-methoxypyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MW: 158.586 g/mol
MDL Number: MFCD09033793
CAS Number: 914222-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR