5,6-Dichloroindolin-2-one 98%

Supplier: Ambeed
Warning

A931729-1G A931729-250MG A931729-5G
76776-994EA 50.93 USD
76776-994 76776-996 77595-508
5,6-Dichloroindolin-2-one 98%
5,6-Dichloroindolin-2-one

Precaución: for Research Use Only

Formula: C₈H₅Cl₂NO
MDL Number: MFCD08692699
CAS Number: 71293-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR