6-Methoxypyridazine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A931185-100MG A931185-1G A931185-250MG A931185-5G
76776-990EA 26 USD
76776-990 76776-992 77603-314 77878-743
6-Methoxypyridazine-3-carboxylic acid 95%
6-Methoxypyridazine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MDL Number: MFCD09800587
CAS Number: 56434-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR