2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone

A930889-100G A930889-10G A930889-25G A930889-500G A930889-5G
76776-980EA 35.01 USD
76776-980 76776-982 76776-984 76776-986 76776-988
2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone 98%
2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone

Precaución: for Research Use Only

Formula: C₁₀H₇F₂N₃O
MW: 223.18 g/mol
MDL Number: MFCD02093825
CAS Number: 86404-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR