2-Amino-3-chloro-5-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A929899-100G A929899-25G A929899-500G A929899-5G
76776-972EA 54.99 USD
76776-972 76776-974 76776-976 76776-978
2-Amino-3-chloro-5-nitrobenzonitrile 98%
2-Amino-3-chloro-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MDL Number: MFCD00126957
CAS Number: 20352-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR