6-Chloro-7-nitro-2H-1,4-benzoxazin-3(4H)-one 95%

Supplier: Ambeed
Danger

Synonyms: 6-Chloro-7-nitro-2H-benzo[b][1,4]oxazin-3(4H)-one

A929051-1G A929051-250MG A929051-5G
76776-966EA 18 USD
76776-966 76776-968 76776-970
6-Chloro-7-nitro-2H-1,4-benzoxazin-3(4H)-one 95%
6-Chloro-7-nitro-2H-1,4-benzoxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₄
MW: 228.59 g/mol
MDL Number: MFCD02660643
CAS Number: 116862-22-7
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR