4-Pyridoxic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxylic acid, 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinic acid

A919047-1G A919047-100MG A919047-250MG
76776-960EA 450 USD
76776-960 77878-739 77878-740
4-Pyridoxic acid 97%
4-Pyridoxic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MW: 183.2 g/mol
MDL Number: MFCD00006334
CAS Number: 82-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR