1-(2-Bromoethyl)-3-(trifluoromethyl)benzene 95%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-2-[3-(trifluoromethyl)phenyl]ethane

A915348-250MG A915348-5G
76776-958EA 43.5 USD
76776-958 77627-748
1-(2-Bromoethyl)-3-(trifluoromethyl)benzene 95%
1-(2-Bromoethyl)-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈BrF₃
MW: 253.062 g/mol
MDL Number: MFCD03092900
CAS Number: 1997-80-4
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR