2-Fluoro-6-(trifluoromethyl)benzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2-fluoro-6-(trifluoromethyl)benzene, 2-(Bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene

A914595-10G A914595-1G A914595-250MG A914595-25G A914595-5G
76776-950EA 66.84 USD
76776-950 76776-952 76776-954 76776-956 77627-746
2-Fluoro-6-(trifluoromethyl)benzyl bromide 98%
2-Fluoro-6-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₅BrF₄
MW: 257.03 g/mol
MDL Number: MFCD00082477
CAS Number: 239087-08-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR