3-Hydroxy-2,4,6-tribromobenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,4,6-Tribromo-3-hydroxybenzoic acid

A912382-100G A912382-1G A912382-25G A912382-5G
76776-942EA 375.56 USD
76776-942 76776-944 76776-946 76776-948
3-Hydroxy-2,4,6-tribromobenzoic acid 98%
3-Hydroxy-2,4,6-tribromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Br₃O₃
MW: 374.81 g/mol
MDL Number: MFCD00055557
CAS Number: 14348-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR