5-Fluoro-2-methyl-1H-indene-3-acetic acid 98%

Supplier: Ambeed
Warning

A909412-1G A909412-25G A909412-250MG
76776-938EA 36 USD
76776-938 76776-940 77878-727
5-Fluoro-2-methyl-1H-indene-3-acetic acid 98%
5-Fluoro-2-methyl-1H-indene-3-acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁FO₂
MDL Number: MFCD01076194
CAS Number: 32004-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR