5-Fluoro-2-iodobenzonitrile 95%

Supplier: Ambeed
Warning

A908077-1G A908077-250MG A908077-5G
76776-936EA 47 USD
76776-936 77878-725 77878-726
5-Fluoro-2-iodobenzonitrile 95%
5-Fluoro-2-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FIN
MDL Number: MFCD07782077
CAS Number: 877868-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR