(5-Methylthiophen-2-yl)methanamine 95%

Supplier: Ambeed
Danger

A895409-250MG A895409-5G A895409-1G
76776-922EA 33.95 USD
76776-922 76776-924 77603-238
(5-Methylthiophen-2-yl)methanamine 95%
(5-Methylthiophen-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₉NS
MW: 127.21 g/mol
MDL Number: MFCD00965306
CAS Number: 104163-34-0
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR