6-(Trifluoromethyl)-1H-benzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 6-(Trifluoromethyl)-1H-benzo[d]imidazole

A887185-1G A887185-250MG A887185-5G
76776-910EA 81.69 USD
76776-910 76776-912 77603-222
6-(Trifluoromethyl)-1H-benzimidazole 98%
6-(Trifluoromethyl)-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂
MW: 186.134 g/mol
MDL Number: MFCD09027453
CAS Number: 326-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR