Rubidium chloride 99%

Supplier: Ambeed
Warning

Synonyms: Rubidiumchloride

A887101-100G A887101-5G A887101-25G
76776-898EA 262 USD
76776-898 76776-900 77595-434
Rubidium chloride 99%
Rubidium chloride

Precaución: for Research Use Only

Formula: RbCl
MW: 120.92 g/mol
Boiling Pt: 1390
Melting Pt: 715
MDL Number: MFCD00011187
CAS Number: 7791-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR