4-Bromo-3-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2-bromobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-m-toluidine, 5-Amino-2-bromo-α,α,α-trifluorotoluene

A884083-100G A884083-10G A884083-1G A884083-25G A884083-500G A884083-5G A884083-1000G
76776-880EA 40.32 USD
76776-880 76776-882 76776-884 76776-886 76776-888 76776-890 77627-710
4-Bromo-3-(trifluoromethyl)aniline 98%
4-Bromo-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
MDL Number: MFCD00007827
CAS Number: 393-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR