Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-asparagine 96%

Supplier: Ambeed
Warning

Synonyms: Nɑ-Fmoc-D-asparagine, Fmoc-D-Asn-OH, Fmoc-D-Asparagine

A884062-10G A884062-1G A884062-25G A884062-100G A884062-5G
76776-874EA 18 USD
76776-874 76776-876 76776-878 77627-706 77627-708
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-asparagine 96%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-asparagine

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₂O₅
MDL Number: MFCD00062956
CAS Number: 108321-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR