5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 98%

Supplier: Ambeed
Warning

A883399-1G A883399-250MG A883399-25G A883399-5G
76776-866EA 50.93 USD
76776-866 76776-868 76776-870 76776-872
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 98%
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂O
MW: 212.56 g/mol
MDL Number: MFCD00068130
CAS Number: 128455-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR