2-(3-(Carboxymethyl)-4-(phenylthio)phenyl)propanoic acid 99%

Supplier: Ambeed
Warning

A883305-25G A883305-5G
76776-862EA 58.35 USD
76776-862 76776-864
2-(3-(Carboxymethyl)-4-(phenylthio)phenyl)propanoic acid 99%
2-(3-(Carboxymethyl)-4-(phenylthio)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₄S
MW: 316.37 g/mol
MDL Number: MFCD09952329
CAS Number: 83237-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR