6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one 95%

Supplier: Ambeed
Danger

A873632-1G A873632-10G A873632-250MG A873632-5G
76776-856EA 37 USD
76776-856 77627-698 77878-711 77878-712
6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one 95%
6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₃ClN₄O
MDL Number: MFCD01219639
CAS Number: 33050-32-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR