ω-Chloro-2,4-difluoroacetophenone 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-1-(2,4-difluorophenyl)ethanone, α-Chloro-2,4-difluoroacetophenone, 2-Chloro-2',4'-difluoroacetophenone

A872923-10G A872923-25G A872923-5G
76776-842EA 10.61 USD
76776-842 76776-844 76776-846
ω-Chloro-2,4-difluoroacetophenone 98%
ω-Chloro-2,4-difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O
MW: 190.58 g/mol
MDL Number: MFCD00013252
CAS Number: 51336-94-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR