α,α,α-Trifluoro-p-toluidine 98%

Supplier: Ambeed
Danger

A871006-10G A871006-25G A871006-100G
76776-838EA 5 USD
76776-838 76776-840 77599-198
α,α,α-Trifluoro-p-toluidine 98%
α,α,α-Trifluoro-p-toluidine

Precaución: for Research Use Only

Formula: C₇H₆F₃N
MDL Number: MFCD00064396
CAS Number: 455-14-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR