4-Amino-4'-nitrodiphenyl sulfide 98%

Supplier: Ambeed
Warning

Synonyms: 4-Aminophenyl 4-Nitrophenyl sulphide, 4-((4-Nitrophenyl)thio)aniline, 4-(4-Nitrophenylthio)aniline

A870039-25G A870039-5G A870039-1G A870039-100G
76776-834EA 84 USD
76776-834 76776-836 77627-692 77878-710
4-Amino-4'-nitrodiphenyl sulfide 98%
4-Amino-4'-nitrodiphenyl sulfide

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂S
MW: 246.28 g/mol
MDL Number: MFCD00007881
CAS Number: 101-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR