(3,4,5-Trimethoxyphenyl)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3,4,5-Trimethoxyphenylacetonitrile

A868877-10G A868877-1G A868877-25G A868877-5G
76776-820EA 53.05 USD
76776-820 76776-822 76776-824 77599-196
(3,4,5-Trimethoxyphenyl)acetonitrile 97%
(3,4,5-Trimethoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
MDL Number: MFCD00001912
CAS Number: 13338-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR