5-Phenyl-1,3-oxazol-2-amine 96%

Supplier: Ambeed
Warning

Synonyms: 5-Phenyloxazol-2-amine

A865995-100MG A865995-250MG A865995-1G
76776-810EA 67 USD
76776-810 76776-812 77573-750
5-Phenyl-1,3-oxazol-2-amine 96%
5-Phenyl-1,3-oxazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD00087876
CAS Number: 6826-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR