o,o,o-Tritolyl phosphate 97%

Supplier: Ambeed
Danger

Synonyms: Tri-o-tolyl phosphate, o,o,o-Tricresyl phosphate, Tris(2-methylphenyl) phosphate, o,o,o-TCP

A862398-25G A862398-5G A862398-100G
76776-786EA 19 USD
76776-786 76776-788 77569-600
o,o,o-Tritolyl phosphate 97%
o,o,o-Tritolyl phosphate

Precaución: For research use only.

Formula: C₂₁H₂₁O₄P
MW: 368.37 g/mol
Boiling Pt: 410
Melting Pt: 11
MDL Number: MFCD00041906
CAS Number: 78-30-8
UN: 3082
ADR: 9 Ⅲ
Merck Index: 12,09893

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR