5-Amino-1,4-benzodioxane 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Benzodioxan-5-amine, 2,3-Dihydrobenzo[b][1,4]dioxin-5-amine

A860846-1G A860846-250MG A860846-25G A860846-5G
76776-778EA 38 USD
76776-778 76776-780 76776-782 76776-784
5-Amino-1,4-benzodioxane 98%
5-Amino-1,4-benzodioxane

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.1626 g/mol
MDL Number: MFCD03695459
CAS Number: 16081-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR