(S)-(+)-2-Octanol 98%

Supplier: Ambeed
Warning

Synonyms: D(+)-2-Octanol, (S)-Octan-2-ol, (S)-(+)-Hexyl methyl carbinol, (S)-(+)-2-Hydroxy octane, (S)-(+)-Methyl hexyl carbinol

A859468-100G A859468-1G A859468-25G A859468-5G
76776-770EA 443.46 USD
76776-770 76776-772 76776-774 76776-776
(S)-(+)-2-Octanol 98%
(S)-(+)-2-Octanol

Precaución: for Research Use Only

Formula: C₈H₁₈O
MW: 130.23 g/mol
MDL Number: MFCD00064283
CAS Number: 6169-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR