5-Bromo-2-(2H-tetrazol-5-yl)pyridine 95%

Supplier: Ambeed
Danger

A858339-250MG A858339-5G A858339-1G
76776-766EA 43.5 USD
76776-766 76776-768 77627-670
5-Bromo-2-(2H-tetrazol-5-yl)pyridine 95%
5-Bromo-2-(2H-tetrazol-5-yl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₅
MW: 226.03 g/mol
MDL Number: MFCD08062726
CAS Number: 380380-60-9
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR