5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine 96%

Supplier: Ambeed
Warning

A857207-100MG A857207-1G A857207-250MG A857207-25G A857207-5G
76776-756EA 7 USD
76776-756 76776-758 76776-760 76776-762 76776-764
5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine 96%
5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine

Precaución: for Research Use Only

Formula: C₇H₆N₄S
MDL Number: MFCD00473692
CAS Number: 2002-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR