5-Hydroxybenzo[b]furan 98%

Supplier: Ambeed
Warning

A855995-250MG A855995-100MG
76776-754EA 55 USD
76776-754 77878-702
5-Hydroxybenzo[b]furan 98%
5-Hydroxybenzo[b]furan

Precaución: for Research Use Only

Formula: C₈H₆O₂
MDL Number: MFCD10699412
CAS Number: 13196-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR