4-Bromo-2-(trifluoromethoxy)aniline 98%

Supplier: Ambeed
Danger

A855674-10G A855674-25G
76776-748EA 9.55 USD
76776-748 76776-750
4-Bromo-2-(trifluoromethoxy)aniline 98%
4-Bromo-2-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
MDL Number: MFCD00179338
CAS Number: 175278-09-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR