5-Chloro-1-pentyne 96%

Supplier: Ambeed
Danger

A854316-1G A854316-25G A854316-5G
76776-742EA 6.37 USD
76776-742 76776-744 76776-746
5-Chloro-1-pentyne 96%
5-Chloro-1-pentyne

Precaución: for Research Use Only

Formula: C₅H₇Cl
MW: 102.56 g/mol
MDL Number: MFCD00001014
CAS Number: 14267-92-6
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR