5-Tridecanol 98%

Supplier: Ambeed
Warning

A854138-1G A854138-250MG
76776-738EA 477.41 USD
76776-738 76776-740
5-Tridecanol 98%
5-Tridecanol

Precaución: for Research Use Only

Formula: C₁₃H₂₈O
MW: 200.3608 g/mol
MDL Number: MFCD00046731
CAS Number: 58783-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR