Ethyl hydrogen maleate 95%

Supplier: Ambeed
Warning

Synonyms: Monoethyl Maleate, Maleic Acid Monoethyl Ester

A850528-25G A850528-5G A850528-100G
76776-732EA 24 USD
76776-732 76776-734 77599-176
Ethyl hydrogen maleate 95%
Ethyl hydrogen maleate

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.13 g/mol
MDL Number: MFCD00063175
CAS Number: 3990-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR