4,7-Dichloroisatin 95%

Supplier: Ambeed
Warning

Synonyms: 4,7-Dichloroindoline-2,3-dione, 4,7-Dichloro-1H-indole-2,3-dione

A849061-1G A849061-250MG A849061-25G A849061-5G
76776-720EA 11.67 USD
76776-720 76776-722 76776-724 76776-726
4,7-Dichloroisatin 95%
4,7-Dichloroisatin

Precaución: for Research Use Only

Formula: C₈H₃Cl₂NO₂
MW: 216.02 g/mol
MDL Number: MFCD00047214
CAS Number: 18711-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR