3-Cyano-4-fluorobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A842649-1G A842649-5G
76776-708EA 15 USD
76776-708 76776-710
3-Cyano-4-fluorobenzenesulfonyl chloride 98%
3-Cyano-4-fluorobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃ClFNO₂S
MDL Number: MFCD03093760
CAS Number: 351003-23-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR