5-Hydroxyflavone 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-2-phenyl-4H-chromen-4-one, 5-Hydroxy-2-phenyl-4-benzopyrone, 5-Hydroxy-2-phenylchromen-4-one, 5-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 5-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one

A842385-100MG A842385-1G A842385-250MG A842385-5G
76776-702EA 19.1 USD
76776-702 76776-704 76776-706 77573-680
5-Hydroxyflavone 98%
5-Hydroxyflavone

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₃
MW: 238.24 g/mol
MDL Number: MFCD00016944
CAS Number: 491-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR