3-(Oxiran-2-yl)-7-oxabicyclo[4.1.0]heptane 95%

Supplier: Ambeed
Danger

A841622-10G A841622-1G A841622-250MG A841622-5G A841622-25G
76776-694EA 125 USD
76776-694 76776-696 76776-698 76776-700 77599-160
3-(Oxiran-2-yl)-7-oxabicyclo[4.1.0]heptane 95%
3-(Oxiran-2-yl)-7-oxabicyclo[4.1.0]heptane

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MDL Number: MFCD00022354
CAS Number: 106-87-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR