5,10,15,20-Tetra(4-pyridyl)porphyrin 95%

Supplier: Ambeed
Warning

Synonyms: 5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine, meso-Tetra(4-pyridyl)porphine

A841175-1G A841175-250MG A841175-5G A841175-100MG
76776-686EA 52 USD
76776-686 76776-688 76776-690 77878-689
5,10,15,20-Tetra(4-pyridyl)porphyrin 95%
5,10,15,20-Tetra(4-pyridyl)porphyrin

Precaución: for Research Use Only

Formula: C₄₀H₂₆N₈
MW: 618.69 g/mol
MDL Number: MFCD00009626
CAS Number: 16834-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR