2,3-Difluorobenzonitrile 96%

Supplier: Ambeed
Warning

A830861-100G A830861-1G A830861-25G A830861-5G
76776-648EA 318 USD
76776-648 76776-650 76776-652 76776-654
2,3-Difluorobenzonitrile 96%
2,3-Difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃F₂N
MDL Number: MFCD00009976
CAS Number: 21524-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR