α-Methyl-2-trifluoromethylbenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 1-[2-(Trifluoromethyl)phenyl]ethanol

A829377-100G A829377-10G A829377-1G A829377-25G A829377-5G
76776-638EA 211 USD
76776-638 76776-640 76776-642 76776-644 76776-646
α-Methyl-2-trifluoromethylbenzyl alcohol 97%
alpha-Methyl-2-trifluoromethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₉F₃O
MDL Number: MFCD00004513
CAS Number: 79756-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR