3-(6-Chloro-3-oxo-2,3-dihydrobenzo[1,4]oxazin-4-yl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(6-Chloro-3-oxo-2H-benzo[b][1,4]oxazin-4(3H)-yl)propanoic acid

A828873-1G A828873-25G A828873-5G
76776-632EA 12.74 USD
76776-632 76776-634 76776-636
3-(6-Chloro-3-oxo-2,3-dihydrobenzo[1,4]oxazin-4-yl)propionic acid 98%
3-(6-Chloro-3-oxo-2,3-dihydrobenzo[1,4]oxazin-4-yl)propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClNO₄
MW: 255.66 g/mol
MDL Number: MFCD03424566
CAS Number: 351003-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR